CAS RN 447-31-4
CAS RN 447-31-4
IUPAC Name:
2-chloro-1,2-diphenylethan-1-one
Synonyms:
2-chloro-1,2-diphenylethanone
Molecular Weight (g/mol):
230.69
Molecular Formula:
C14H11ClO
InChi Key:
RXDYOLRABMJTEF-UHFFFAOYNA-N
SMILES:
ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Desyl chloride, 98%
CAS: 447-31-4 Molecular Formula: C14H11ClO Molecular Weight (g/mol): 230.69 MDL Number: MFCD00000858 InChI Key: RXDYOLRABMJTEF-UHFFFAOYNA-N Synonym: desyl chloride,2-chloro-2-phenylacetophenone,alpha-chlorodeoxybenzoin,1,2-diphenyl-2-chloroethanone,ethanone, 2-chloro-1,2-diphenyl,alpha-chlorobenzyl phenyl ketone,2-chloro-1,2-diphenylethone,acetophenone, alpha-chloro-alpha-phenyl,2-chloro-1,2-diphenyl-ethanone,acetophenone, 2-chloro-2-phenyl PubChem CID: 95343 IUPAC Name: 2-chloro-1,2-diphenylethanone SMILES: ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1