accessibility menu, dialog, popup

UserName

CAS RN 4469-80-1

H3CONH2

CAS RN 4469-80-1

IUPAC Name: 4-propoxyaniline
Synonyms: 4-propoxyaniline
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: DWOIGSLSPPLRKO-UHFFFAOYSA-N
SMILES: CCCOC1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results