accessibility menu, dialog, popup

UserName

CAS RN 444932-31-4

H3COOHNNHONHOCH3

CAS RN 444932-31-4

IUPAC Name: 3,5-dimethoxy-N-{[(naphthalen-1-yl)carbamoyl]amino}benzamide
Synonyms: 3,5-dimethoxy-N-{[(naphthalen-1-yl)carbamoyl]amino}benzamide
Molecular Weight (g/mol): 365.39
Molecular Formula: C20H19N3O4
InChi Key: KPTMSQHTGZMEFU-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C(=O)NNC(=O)NC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results