accessibility menu, dialog, popup

UserName

CAS RN 4439-20-7

HOHNNHOH

CAS RN 4439-20-7

IUPAC Name: 2-({2-[(2-hydroxyethyl)amino]ethyl}amino)ethan-1-ol
Synonyms: 2-({2-[(2-hydroxyethyl)amino]ethyl}amino)ethanol
Molecular Weight (g/mol): 148.21
Molecular Formula: C6H16N2O2
InChi Key: GFIWSSUBVYLTRF-UHFFFAOYSA-N
SMILES: OCCNCCNCCO
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results