accessibility menu, dialog, popup

UserName

H2NHNOHOOO(S)OHNH(S)HOOHHO

CAS RN 4368-28-9

IUPAC Name: (11S,13S)-3-amino-5,12,13,14-tetrahydroxy-14-(hydroxymethyl)-8,10-dioxa-2,4-diazatetracyclo[7.3.1.1⁷,¹¹.0¹,⁶]tetradec-3-en-4-ium-9-olate
Synonyms: tetrodotoxin
Molecular Weight (g/mol): 319.27
Molecular Formula: C11H17N3O8
InChi Key: SLBCPBUVHCASIJ-UFHSVNPDSA-O
SMILES: NC1=[NH+]C(O)C2C3OC4([O-])O[C@@H](C(O)C2(N1)[C@@H]4O)C3(O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results