accessibility menu, dialog, popup

UserName

FFFBrN

CAS RN 436799-32-5

IUPAC Name: 5-bromo-2-(trifluoromethyl)pyridine
Synonyms: 5-bromo-2-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 226
Molecular Formula: C6H3BrF3N
InChi Key: RPFAUCIXZGMCFN-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(Br)C=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results