CAS RN 4337-43-3
CAS RN 4337-43-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-Chloro-1,2-benzisothiazol-3(2H)-one, TRC
CAS: 4337-43-3 Molecular Formula: C7 H4 Cl N O S Molecular Weight (g/mol): 185.63 IUPAC Name: 5-chloro-1,2-benzothiazol-3-one SMILES: Clc1ccc2SNC(=O)c2c1