accessibility menu, dialog, popup

UserName

CAS RN 4334-88-7

IUPAC Name: [4-(ethoxycarbonyl)phenyl]boronic acid
Synonyms: 4-(ethoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 193.99
Molecular Formula: C9H11BO4
InChi Key: ZLNFACCFYUFTLD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(C=C1)B(O)O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results