accessibility menu, dialog, popup

UserName

FFFOBrBr

CAS RN 431-67-4

IUPAC Name: 3,3-dibromo-1,1,1-trifluoropropan-2-one
Synonyms: 3,3-dibromo-1,1,1-trifluoropropan-2-one
Molecular Weight (g/mol): 269.84
Molecular Formula: C3HBr2F3O
InChi Key: HEPPAPZASXFWTB-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C(Br)Br
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)