CAS RN 431-34-5
CAS RN 431-34-5
IUPAC Name:
3-bromo-1,1,1-trifluoropropan-2-ol
Synonyms:
3-bromo-1,1,1-trifluoropropan-2-ol
Molecular Weight (g/mol):
192.96
Molecular Formula:
C3H4BrF3O
InChi Key:
VBHIIZIQRDVGDH-UHFFFAOYNA-N
SMILES:
OC(CBr)C(F)(F)F
1
–
1
of
1
results
1
3-Bromo-1,1,1-trifluoro-2-propanol, 96%
CAS: 431-34-5 Molecular Formula: C3H4BrF3O Molecular Weight (g/mol): 192.96 MDL Number: MFCD00040945 InChI Key: VBHIIZIQRDVGDH-UHFFFAOYNA-N PubChem CID: 220632 IUPAC Name: 3-bromo-1,1,1-trifluoropropan-2-ol SMILES: OC(CBr)C(F)(F)F