accessibility menu, dialog, popup

UserName

CAS RN 4292-92-6

H3C

CAS RN 4292-92-6

IUPAC Name: pentylcyclohexane
Synonyms: pentylcyclohexane
Molecular Weight (g/mol): 154.3
Molecular Formula: C11H22
InChi Key: HLTMUYBTNSVOFY-UHFFFAOYSA-N
SMILES: CCCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results