accessibility menu, dialog, popup

UserName

CAS RN 42877-08-7

H3COSBr

CAS RN 42877-08-7

IUPAC Name: 1-(3-bromothiophen-2-yl)ethan-1-one
Synonyms: 1-(3-bromothiophen-2-yl)ethanone
Molecular Weight (g/mol): 205.07
Molecular Formula: C6H5BrOS
InChi Key: ZUXPELAHJQSZTE-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Br)C=CS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results