accessibility menu, dialog, popup

UserName

H3CCH3H3COONCH3OOCH3

CAS RN 42492-57-9

IUPAC Name: methyl 2-{[(tert-butoxy)carbonyl](methyl)amino}acetate
Synonyms: methyl 2-[(tert-butoxycarbonyl)(methyl)amino]acetate
Molecular Weight (g/mol): 203.24
Molecular Formula: C9H17NO4
InChi Key: LTBDTYJYKRLTRN-UHFFFAOYSA-N
SMILES: COC(=O)CN(C)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)