CAS RN 4249-72-3
CAS RN 4249-72-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Phenoxy-1-phenylethanol, TRC
CAS: 4249-72-3 Molecular Formula: C14H14O2 Molecular Weight (g/mol): 214.25 Synonym: alpha-(Phenoxymethyl)benzenemethanol,-Phenyl-2-phenoxyethanol,2-Phenoxy-1-phenethanol,2-Phenoxy-1-phenylethan-1-ol,2-Phenoxy-1-phenylethanol IUPAC Name: 2-phenoxy-1-phenylethanol SMILES: OC(COc1ccccc1)c2ccccc2