accessibility menu, dialog, popup

UserName

CAS RN 421595-78-0

NNONHNH2

CAS RN 421595-78-0

IUPAC Name: imidazo[1,2-a]pyridine-7-carbohydrazide
Synonyms: imidazo[1,2-a]pyridine-7-carbohydrazide
Molecular Weight (g/mol): 176.18
Molecular Formula: C8H8N4O
InChi Key: VXJBLZXLLRYHPD-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC2=NC=CN2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results