accessibility menu, dialog, popup

UserName

FFClFFI

CAS RN 421-78-3

IUPAC Name: 1-chloro-1,1,2,2-tetrafluoro-2-iodoethane
Synonyms: 1-chloro-1,1,2,2-tetrafluoro-2-iodoethane
Molecular Weight (g/mol): 262.37
Molecular Formula: C2ClF4I
InChi Key: XNIBSTIKPHNKSO-UHFFFAOYSA-N
SMILES: FC(F)(Cl)C(F)(F)I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results