accessibility menu, dialog, popup

UserName

CH3NNSCH3Na

CAS RN 41840-27-1

IUPAC Name: sodium (4,6-dimethylpyrimidin-2-yl)sulfanide
Synonyms: sodium (4,6-dimethylpyrimidin-2-yl)sulfanide
Molecular Weight (g/mol): 162.19
Molecular Formula: C6H7N2NaS
InChi Key: HUDKHPCVAMVYEQ-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC(C)=NC([S-])=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results