accessibility menu, dialog, popup

UserName

CH3NSBr

CAS RN 41731-23-1

IUPAC Name: 2-bromo-5-methyl-1,3-thiazole
Synonyms: 2-bromo-5-methyl-1,3-thiazole
Molecular Weight (g/mol): 178.05
Molecular Formula: C4H4BrNS
InChi Key: FJPZHYAYNAUKKA-UHFFFAOYSA-N
SMILES: CC1=CN=C(Br)S1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)