accessibility menu, dialog, popup

UserName

CAS RN 41288-96-4

OHClN

CAS RN 41288-96-4

IUPAC Name: 6-chloropyridin-3-ol
Synonyms: 6-chloropyridin-3-ol
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: KVCOOWROABTXDJ-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results