accessibility menu, dialog, popup

UserName

CAS RN 41036-32-2

H3C(R)NH2OOCH3SOHOO

CAS RN 41036-32-2

IUPAC Name: 4-methylbenzene-1-sulfonic acid; benzyl (2R)-2-aminopropanoate
Synonyms: benzyl (2R)-2-aminopropanoate; para-toluene sulfonate
Molecular Weight (g/mol): 351.42
Molecular Formula: C17H21NO5S
InChi Key: NWOPHJSSBMABBD-DDWIOCJRSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.C[C@@H](N)C(=O)OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results