accessibility menu, dialog, popup

UserName

HNOOHOH

CAS RN 40990-52-1

IUPAC Name: 4-hydroxy-1H-indole-6-carboxylic acid
Synonyms: 4-hydroxy-1H-indole-6-carboxylic acid
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: TXZOAKBAZXCPGT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(O)=C2C=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results