accessibility menu, dialog, popup

UserName

CH3OOHOOCH3CH3O

CAS RN 4099-85-8

IUPAC Name: {6-methoxy-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}methanol
Synonyms: {6-methoxy-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-yl}methanol
Molecular Weight (g/mol): 204.22
Molecular Formula: C9H16O5
InChi Key: DXBHDBLZPXQALN-UHFFFAOYNA-N
SMILES: COC1OC(CO)C2OC(C)(C)OC12
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (6)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)