accessibility menu, dialog, popup

UserName

FNCS

CAS RN 404-72-8

IUPAC Name: 1-fluoro-3-isothiocyanatobenzene
Synonyms: 1-fluoro-3-isothiocyanatobenzene
Molecular Weight (g/mol): 153.17
Molecular Formula: C7H4FNS
InChi Key: HDBACITVPQEAGG-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results