accessibility menu, dialog, popup

UserName

CAS RN 40397-98-6

IUPAC Name: 2-isocyanato-1,4-dimethylbenzene
Synonyms: 2,5-dimethylphenyl isocyanate 2,5-dimethyl phenylisocyanate
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: SOXVXJQIQVOCAY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C)C(=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results