accessibility menu, dialog, popup

UserName

HOONN

CAS RN 40332-16-9

IUPAC Name: 2-(1H-1,3-benzodiazol-1-yl)acetic acid
Synonyms: 1,3-benzodiazol-1-ylacetic acid
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: AFHSZBKDUTWXDX-UHFFFAOYSA-N
SMILES: OC(=O)CN1C=NC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results