CAS RN 402849-27-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-2-(4-chlorobenzoyl)-1-(4-methoxyphenyl)hydrazide Benzoic Acid, TRC
CAS: 402849-27-8 Molecular Formula: C21H16Cl2N2O3 Molecular Weight (g/mol): 415.27 Synonym: Benzoic acid, 4-chloro-, 2-(4-chlorobenzoyl)-1-(4-methoxyphenyl)hydrazide,4-Chloro-N'-(4-chlorobenzoyl)-N-(4-methoxyphenyl)benzohydrazide,Indometacin dibenzylate IUPAC Name: 4-chloro-N'-(4-chlorobenzoyl)-N'-(4-methoxyphenyl)benzohydrazide SMILES: COc1ccc(cc1)N(NC(=O)c2ccc(Cl)cc2)C(=O)c3ccc(Cl)cc3