accessibility menu, dialog, popup

UserName

H2NBr

CAS RN 3959-07-7

IUPAC Name: 1-(4-bromophenyl)methanamine
Synonyms: 4-bromo-benzylamine
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: XRNVSPDQTPVECU-UHFFFAOYSA-N
SMILES: NCC1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results