accessibility menu, dialog, popup

UserName

CAS RN 395-33-5

OHOHOF

CAS RN 395-33-5

IUPAC Name: 2-(4-fluorophenyl)-2-hydroxyacetic acid
Synonyms: (4-fluorophenyl)(hydroxy)acetic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: RWCMOQXHIDWDDJ-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results