accessibility menu, dialog, popup

UserName

CAS RN 394-47-8

FN

CAS RN 394-47-8

IUPAC Name: 2-fluorobenzonitrile
Synonyms: 2-fluorobenzonitrile
Molecular Weight (g/mol): 121.11
Molecular Formula: C7H4FN
InChi Key: GDHXJNRAJRCGMX-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1C#N
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (3)
  • (3)