accessibility menu, dialog, popup

UserName

OOHSH

CAS RN 39161-84-7

IUPAC Name: 2-(4-sulfanylphenyl)acetic acid
Synonyms: 4-mercaptophenylacetic acid
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: ORXSLDYRYTVAPC-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(S)C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)