accessibility menu, dialog, popup

UserName

ClOS

CAS RN 39098-97-0

IUPAC Name: 2-(thiophen-2-yl)acetyl chloride
Synonyms: 2-(thiophen-2-yl)acetyl chloride
Molecular Weight (g/mol): 160.62
Molecular Formula: C6H5ClOS
InChi Key: AJYXPNIENRLELY-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC=CS1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)