accessibility menu, dialog, popup

UserName

CAS RN 38786-67-3

CH3(S)BrOBr

CAS RN 38786-67-3

IUPAC Name: (2S)-2-bromo-1-(4-bromophenyl)propan-1-one
Synonyms: (2S)-2-bromo-1-(4-bromophenyl)propan-1-one
Molecular Weight (g/mol): 291.97
Molecular Formula: C9H8Br2O
InChi Key: GKALOSTUZMFUQB-LURJTMIESA-N
SMILES: C[C@H](Br)C(=O)C1=CC=C(Br)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results