CAS RN 38613-77-3
CAS RN 38613-77-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tetrakis(2,4-di-tert-butylphenyl)[1,1'-biphenyl]-4,4'-diylbisphosphonite (Technical Grade), TRC
CAS: 38613-77-3 Molecular Formula: C68 H92 O4 P2 Molecular Weight (g/mol): 1035.4 Synonym: Tetrakis(2,4-di-tert-butylphenyl-4,4'-biphenylylene-diphosphonite),Tetrakis(2,4-di-tert-butylphenyl-4,4'-biphenylylene-diphosphonite),Hostanox P-EPQ,Sandostab EPQ IUPAC Name: [4-[4-bis(2,4-ditert-butylphenoxy)phosphanylphenyl]phenyl]-bis(2,4-ditert-butylphenoxy)phosphane SMILES: CC(C)(C)c1ccc(OP(Oc2ccc(cc2C(C)(C)C)C(C)(C)C)c3ccc(cc3)c4ccc(cc4)P(Oc5ccc(cc5C(C)(C)C)C(C)(C)C)Oc6ccc(cc6C(C)(C)C)C(C)(C)C)c(c1)C(C)(C)C