accessibility menu, dialog, popup

UserName

CAS RN 38445-23-7

H3COO

CAS RN 38445-23-7

IUPAC Name: 6-methyl-4H-chromen-4-one
Synonyms: 6-methylchromen-4-one
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: HTXQVFXXVXOLCF-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC=CC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results