CAS RN 380560-89-4
CAS RN 380560-89-4
IUPAC Name:
4-[5-(4-bromophenyl)-3-(6-methyl-2-oxo-4-phenyl-2,3-dihydroquinolin-3-ylidene)pyrazolidin-1-yl]-4-oxobutanoic acid
Synonyms:
4-[5-(4-bromophenyl)-3-(6-methyl-2-oxo-4-phenylquinolin-3-ylidene)pyrazolidin-1-yl]-4-oxobutanoic acid
Molecular Weight (g/mol):
558.43
Molecular Formula:
C29H24BrN3O4
InChi Key:
CADIBCWPGCNUBD-UHFFFAOYNA-N
SMILES:
CC1=CC2=C(C3=CC=CC=C3)C(=C3CC(N(N3)C(=O)CCC(O)=O)C3=CC=C(Br)C=C3)C(=O)N=C2C=C1
1
–
1
of
1
results
1