CAS RN 38020-81-4
IUPAC Name:
2-iodoacetyl chloride
Synonyms:
2-iodoacetyl chloride
Molecular Weight (g/mol):
204.39
Molecular Formula:
C2H2ClIO
InChi Key:
BSVMPWANOMFSPR-UHFFFAOYSA-N
SMILES:
ClC(=O)CI
1
–
2
of
2
results
1
Iodoacetyl chloride, 97%
CAS: 38020-81-4 Molecular Formula: C2H2ClIO Molecular Weight (g/mol): 204.39 MDL Number: MFCD00013718 InChI Key: BSVMPWANOMFSPR-UHFFFAOYSA-N Synonym: iodoacetyl chloride,iodoacetylchloride,acetyl chloride, iodo,jodacetylchlorid,acetylchloride,iodo,iodoacetic acid chloride,acmc-1aewu,iodoacetyl chloride 5g,acetyl chloride, iodo-6ci,9ci PubChem CID: 3084680 IUPAC Name: 2-iodoacetyl chloride SMILES: ClC(=O)CI