CAS RN 37976-96-8
CAS RN 37976-96-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(3b,7b)-3-(Acetyloxy)-7-hydroxy-androst-5-en-17-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3Beta,7Beta)-3-(Acetyloxy)-7-hydroxy-androst-5-en-17-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3Beta,7Beta)-3-(Acetyloxy)-7-hydroxy-androst-5-en-17-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.