accessibility menu, dialog, popup

UserName

CAS RN 37904-03-3

CH3NHOOHO(E)

CAS RN 37904-03-3

IUPAC Name: (2E)-3-[(4-methylphenyl)carbamoyl]prop-2-enoic acid
Synonyms: (2E)-3-[(4-methylphenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: VLWFSWMZGGZHGD-VOTSOKGWSA-N
SMILES: CC1=CC=C(NC(=O)\C=C\C(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results