CAS RN 3790-45-2
CAS RN 3790-45-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Phenyl-tetrahydrocarboline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.