accessibility menu, dialog, popup

UserName

H3CNH2

CAS RN 3789-60-4

IUPAC Name: 1-phenylbutan-1-amine
Synonyms: 1-phenylbutan-1-amine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: XHOXKVFLASIOJD-UHFFFAOYNA-N
SMILES: CCCC(N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results