CAS RN 37859-42-0
CAS RN 37859-42-0
IUPAC Name:
(1,3-benzothiazol-2-yl)methanol
Synonyms:
1,3-benzothiazol-2-ylmethanol
Molecular Weight (g/mol):
165.21
Molecular Formula:
C8H7NOS
InChi Key:
PQXMQZYDBQBWNL-UHFFFAOYSA-N
SMILES:
OCC1=NC2=CC=CC=C2S1
1
–
2
of
2
results
2
2-Benzothiazolemethanol, 97%
CAS: 37859-42-0 Molecular Formula: C8H7NOS Molecular Weight (g/mol): 165.21 MDL Number: MFCD00226293 InChI Key: PQXMQZYDBQBWNL-UHFFFAOYSA-N Synonym: benzo d thiazol-2-ylmethanol,2-benzothiazolemethanol,2-hydroxymethylbenzothiazole,1,3-benzothiazol-2-yl-methanol,benzothiazol-2-ylmethan-1-ol,zlchem 107,pubchem14535,benzothiazol-2-yl-methanol,chembl71700,1,3-benzothiazol-2-yl methanol PubChem CID: 268122 IUPAC Name: 1,3-benzothiazol-2-ylmethanol SMILES: OCC1=NC2=CC=CC=C2S1