accessibility menu, dialog, popup

UserName

CAS RN 37845-05-9

H3COOH

CAS RN 37845-05-9

IUPAC Name: 3-ethyladamantane-1-carboxylic acid
Synonyms: 3-ethyladamantane-1-carboxylic acid
Molecular Weight (g/mol): 208.3
Molecular Formula: C13H20O2
InChi Key: ZJBLNYRNJSFPQO-UHFFFAOYNA-N
SMILES: CCC12CC3CC(C1)CC(C3)(C2)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)