CAS RN 37742-98-6
CAS RN 37742-98-6
IUPAC Name:
4-bromo-2,2-diphenylbutanoic acid
Synonyms:
4-bromo-2,2-diphenylbutanoic acid
Molecular Weight (g/mol):
319.2
Molecular Formula:
C16H15BrO2
InChi Key:
GFIYIIRFIODLLU-UHFFFAOYSA-N
SMILES:
OC(=O)C(CCBr)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
3
of
3
results
1
4-Bromo-2,2-diphenylbutyric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Bromo-2,2-diphenylbutyric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Bromo-2,2-diphenylbutyric Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.