CAS RN 37716-99-7
CAS RN 37716-99-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
3Alpha,4Beta-Dihydroxy-5Alpha-androstan-17-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3Alpha,4Beta-Dihydroxy-5Alpha-androstan-17-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3alpha,4beta-Dihydroxy-5alpha-androstan-17-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.