CAS RN 37682-72-7
CAS RN 37682-72-7
IUPAC Name:
dihydrogen 2-({[(1S)-4-[(diaminomethylidene)amino]-1-{[(2S)-2-({5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}amino)-3-methylbutanoyl]carbamoyl}butyl]carbamoyl}amino)-3-phenylpropanoic acid dichloride
Synonyms:
dihydrogen 2-({[(1S)-4-[(diaminomethylidene)amino]-1-{[(2S)-2-({5-[(diaminomethylidene)amino]-1-oxopentan-2-yl}amino)-3-methylbutanoyl]carbamoyl}butyl]carbamoyl}amino)-3-phenylpropanoic acid dichloride
Molecular Weight (g/mol):
677.63
Molecular Formula:
C27H46Cl2N10O6
InChi Key:
GXQPAKIVOCLCHD-BXLPLHKWSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CC(C)[C@H](NC(CCCN=C(N)N)C=O)C(=O)NC(=O)[C@H](CCCN=C(N)N)NC(=O)NC(CC1=CC=CC=C1)C(O)=O