accessibility menu, dialog, popup

UserName

CAS RN 37558-16-0

H3COO(R)(R)CH3(S)OH(S)OH(R)OH(S)CH3O(R)H3CH3C(S)OOH3C

CAS RN 37558-16-0

IUPAC Name: (1S,2S,6R,10S,11R,13S,14R,15R)-14-(butanoyloxy)-1,6-dihydroxy-8-(hydroxymethyl)-4,12,12,15-tetramethyl-5-oxotetracyclo[8.5.0.0²,⁶.0¹¹,¹³]pentadeca-3,8-dien-13-yl butanoate
Synonyms: phorbol 12,13 dibutyrate 12,13-dibutyrate, phorbol phorbol 12,13-dibutyrate phorbol-12,13-dibutyrate phorbol 12,13-dibutanoate PDBu
Molecular Weight (g/mol): 504.62
Molecular Formula: C28H40O8
InChi Key: BQJRUJTZSGYBEZ-YVQNUNKESA-N
SMILES: CCCC(=O)O[C@@H]1[C@@H](C)[C@]2(O)[C@@H]3C=C(C)C(=O)[C@@]3(O)CC(CO)=C[C@H]2[C@@H]2C(C)(C)[C@]12OC(=O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results