accessibility menu, dialog, popup

UserName

CAS RN 37526-59-3

NH2SOOOFFF

CAS RN 37526-59-3

IUPAC Name: 2-(trifluoromethoxy)benzene-1-sulfonamide
Synonyms: 2-(trifluoromethoxy)benzenesulfonamide
Molecular Weight (g/mol): 241.18
Molecular Formula: C7H6F3NO3S
InChi Key: HIFGQHGWMTZMOH-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC=C1OC(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results