CAS RN 3752-97-4
CAS RN 3752-97-4
IUPAC Name:
1,4-bis(chloromethyl)-2,5-dimethoxybenzene
Synonyms:
1,4-bis(chloromethyl)-2,5-dimethoxybenzene
Molecular Weight (g/mol):
235.1
Molecular Formula:
C10H12Cl2O2
InChi Key:
GEUWDCJVDLHSNQ-UHFFFAOYSA-N
SMILES:
COC1=CC(CCl)=C(OC)C=C1CCl