accessibility menu, dialog, popup

UserName

FFFFIFFFFF

CAS RN 375-51-9

IUPAC Name: 1,1,1,2,2,3,4,4,4-nonafluoro-3-iodobutane
Synonyms: 1,1,1,2,2,3,4,4,4-nonafluoro-3-iodobutane
Molecular Weight (g/mol): 345.93
Molecular Formula: C4F9I
InChi Key: FBJVLVWUMYWJMY-UHFFFAOYNA-N
SMILES: FC(F)(F)C(F)(F)C(F)(I)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)