accessibility menu, dialog, popup

UserName

CAS RN 37421-04-8

H3CNHO

CAS RN 37421-04-8

IUPAC Name: methyl(2-phenoxyethyl)amine
Synonyms: methyl(2-phenoxyethyl)amine
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: GUMBNOITRNRLCS-UHFFFAOYSA-N
SMILES: CNCCOC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results